Learn More
Synthware™ Liebig Condenser
GREENER_CHOICE
With standard taper outer joint at the top and lower inner drip tip joint
Supplier:  Synthware™ C204100
 View more versions of this product
Catalog No. 31500813
Only null left
Description
Description
  • Joints of same size
Specifications
Chemical Identifiers
Glass
Distillation
RC14, R201402
Liebig Condenser
Distillation Head
None
Specifications
Distillation
None
Glass
24/40
Hose Connection: 10mm O.D.
Distillation Head
RC14, R201402
100 mm
Liebig Condenser
Product Suggestions
Product Suggestions