Learn More
Synthware™ Liebig Condenser
GREENER_CHOICE
With standard taper outer joint at the top and lower inner drip tip joint
Supplier:  Synthware™ C209300
 View more versions of this product
Catalog No. 31500812
Only null left
Description
Description
  • Joints of same size
Specifications
Chemical Identifiers
Glass
Distillation
RC14, R201402
Liebig Condenser
Distillation Head
None
Specifications
RC14, R201402
None
Glass
300 mm
Liebig Condenser
Distillation Head
Distillation
19/22
Hose Connection: 8mm O.D.
Product Suggestions
Product Suggestions