Learn More
Synthware™ Kugelrohr Distilling Bulb
GREENER_CHOICE
Used in distillation of very small amount of distillate at lower temperature with high efficiency
Supplier:  Synthware™ D281005
 View more versions of this product
Catalog No. 31501007
Only null left
Description
Description
  • Bulbs have 14/20 standard taper joints at both ends to connect to other bulbs or to single neck round bottom flasks
Specifications
Chemical Identifiers
14/20
Specifications
80mm
5 mL
14/20
Product Suggestions
Product Suggestions