Learn More
Synthware™ Inlet Adapters with Standard Taper Inner Joint
Synthware inlet adapter with a standard taper inner joint.
$74.46 - $78.79
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Inlet adapter
Quantity 1
Standard Taper Joint No. 24/40
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Price Quantity  
31500579
View Documents
Synthware™
A314075
Tube: 75mm
Each for $74.46
Only null left
31500580
View Documents
Synthware™
A314250
Tube: 250mm
Each for $78.79
Only null left
Description
Description
Design allows introduction of reactants below the surface. The serrated hose connections have an O.D. of 10 mm.
Specifications
Specifications
Organic Synthesis
Inlet adapter
24/40
Borosilicate Glass (3.3)
1
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'