missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Inlet Adapters with Standard Taper Inner Joint

Synthware inlet adapter with a standard taper inner joint.

Supplier:  Synthware™ A314075

 View more versions of this product

Catalog No. 31500579


Only null left
Explore available promotions

Description

Description

Design allows introduction of reactants below the surface. The serrated hose connections have an O.D. of 10 mm.
Specifications

Specifications

Inlet Adapter
Hose O.D.: 10 mm
Borosilicate Glass (3.3)
1
Organic Synthesis
Tube: 75mm
Inlet adapter
24/40
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions