missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Gas Dispersion Tubes, Fritted, 90° Angle

For use with filtration techniques

Supplier:  Synthware™ F283045C

 View more versions of this product

Catalog No. 31501552

Only null left
Explore available promotions

Description

Description

  • 90° angle permits bubbles to be spread over a large area
  • With stem O.D. of 8 mm and Length: 150 mm
Specifications

Specifications

Coarse
30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions