Learn More
Synthware™ Flat Bottom Flask, Single Neck
All sizes are hand blown from tubing to ensure a uniform wall thickness
Supplier:  Synthware™ F122450
 View more versions of this product
Catalog No. 31501131
Only null left
Explore available promotions
Description
Description
  • Flat bottom, single short neck flask with standard taper outer joint
  • Heavy wall design
Specifications
Specifications
Joint: 24/40; Single neck
50 mL
Round
Product Suggestions
Product Suggestions