missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Extraction Thimble, Glass

For use with Soxhlet extractor
Supplier: Synthware™ E113590X
Description
- All-glass extraction thimble permits viewing extraction process and subsequent weighing
- Available in coarse and extra coarse porosities
Chemical Identifiers
| For use with Soxhlet extractor | |
| Borosilicate Glass | |
| 1.5 mm |
Specifications
| For use with Soxhlet extractor | |
| Borosilicate Glass | |
| Fits Extractor Size: Medium | |
| Extra Coarse |
| 35 mm | |
| 1.5 mm | |
| 90 mm |