Learn More
Synthware™ Extraction Thimble, Glass
GREENER_CHOICE
For use with Soxhlet extractor
Supplier:  Synthware™ E112585X
 View more versions of this product
Catalog No. 31501080
Only null left
Description
Description
  • All-glass extraction thimble permits viewing extraction process and subsequent weighing
  • Available in coarse and extra coarse porosities
Specifications
Chemical Identifiers
For use with Soxhlet extractor
Borosilicate Glass
1.5 mm
Specifications
For use with Soxhlet extractor
Borosilicate Glass
Fits Extractor Size: Small
Extra Coarse
25 mm
1.5 mm
85 mm
Product Suggestions
Product Suggestions