Learn More
Synthware™ Evaporation Flasks
For controlled evaporation of foaming solvents such as toluene or xylene on rotary evaporator
$52.36 - $277.21
Specifications
Material Borosilicate Glass (3.3)
Type Evaporating Flask
For Use With (Equipment) Rotary Evaporator
Holds Foaming Solvents
View More Specs
Products 9
Catalog Number Mfr. No. Volume (Metric) Standard Taper Joint Number Center Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Standard Taper Joint Number Center Price Quantity  
31501178
View Documents
Synthware™
F791450
50 mL 14/20
Each for $52.36
Only null left
31501180
View Documents
Synthware™
F794100
100 mL 24/40
Each for $84.96
Only null left
31501183
View Documents
Synthware™
F794250
250 mL 24/40
Each for $134.75
Only null left
31501181
View Documents
Synthware™
F794200
200 mL 24/40
Each for $110.89
Only null left
31501179
View Documents
Synthware™
F791100
100 mL 14/20
Each for $66.39
Only null left
31501182
View Documents
Synthware™
F791250
250 mL 14/20
Each for $97.29
Only null left
31501184
View Documents
Synthware™
F798250
250 mL 29/42
Each for $165.00
Only null left
31501185
View Documents
Synthware™
F794500
500 mL 24/40
Each for $226.82
Only null left
31501186
View Documents
Synthware™
F794000
1 L 24/40
Each for $277.21
Only null left
Description
Description
  • Low profile design sits upright on bench
  • Provides easy product access with spatula
Specifications
Specifications
Borosilicate Glass (3.3)
Evaporating Flask
Rotary Evaporator
Foaming Solvents