Learn More
Synthware™ Evaporation Flasks
For controlled evaporation of foaming solvents such as toluene or xylene on rotary evaporator
Supplier:  Synthware™ F794250
 View more versions of this product
Catalog No. 31501183
Only null left
Description
Description
  • Low profile design sits upright on bench
  • Provides easy product access with spatula
Specifications
Specifications
Borosilicate Glass (3.3)
Rotary Evaporator
24/40
Evaporating Flask
Foaming Solvents
250 mL
Product Suggestions
Product Suggestions