Learn More
Synthware™ Distilling Head, Short Path, Jacketed, Vigreux
Short path distilling head has a close coupled condenser and vacuum connection to offer an extremely short condensate travel path
Supplier:  Synthware™ D371418
Catalog No. 31501023
Only null left
Explore available promotions
Description
Description
  • Thermometer stem and bulb serve as column packing
  • Lower hose connection for vacuum, upper hose connections for cooling lines
  • Used with 25mm immersion thermometer
  • Vacuum jacket and Vigreux indentations improve efficiency
Specifications
Specifications
14/20
Thermometer Joint: 10/18
Product Suggestions
Product Suggestions