Learn More
Synthware™ Distillation Receiver, Dean Stark, Vacuum Jacketed
Dean Stark distillation receiver for easy separation of water from reaction or distillation system
Supplier:  Synthware™ D442410
 View more versions of this product
Catalog No. 31501071
Only null left
Explore available promotions
Description
Description
  • Graduated
  • Uptake tube vacuum jacketed to improve distillation performance
  • Jacket eliminates possibility of condensation or recrystallization of distillate in sidearm
  • With upper outer and lower inner standard taper joints, both of same size
  • PTFE stopcock with 2 mm bore
Specifications
Specifications
95mm
Distillation Receiver
24/40
275mm
10 mL
Product Suggestions
Product Suggestions