Learn More
Synthware™ Distillation Condenser with Removable Hose Connections
GREENER_CHOICE
With indentations at the bottom for supporting packing material
Supplier:  Synthware™ C579110
 View more versions of this product
Catalog No. 31500731
Only null left
Description
Description
  • Jacket permits column to be used as condenser
  • With top outer and lower inner standard taper joints
Specifications
Chemical Identifiers
Distillation Condenser
Specifications
200mm
Distillation Condenser
19/22
Product Suggestions
Product Suggestions