missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Distillation Columns, Vacuum Jacketed
GREENER_CHOICE

Catalog No. 31500722
Change view
Click to view available options
Standard Taper Joint:
14/20
24/40
Height (Metric):
Jacket: 130mm
Jacket: 250mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint Height (Metric)
31500722 24/40 Jacket: 250mm
31500721 14/20 Jacket: 130mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31500722 Supplier Synthware™ Supplier No. C164250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vacuum jacketed distilling column for better performance

  • 12mm I.D. indentations to support packing material
  • Top outer and lower inner standard taper joints of same size

Specifications

Height (Metric) Jacket: 250mm
Type Vacuum Jacketed Distillation Column
Standard Taper Joint 24/40