Learn More
Synthware™ Coiled Condenser, with Removable Hose Connections
With inner coil that coolant is circulated through
Supplier: Synthware™ C524400
Description
- Upper outer joint and lower inner joint of same size
Chemical Identifiers
| Hose connection: 8mm O.D. | |
| Glass | |
| Coil Condenser |
Specifications
| 24/40 | |
| Hose connection: 8mm O.D. | |
| Coil Condenser |
| Glass | |
| 400 mm |