Learn More
Synthware™ Chromatography Column with Standard Taper Joint, Reservoir, PTFE Stopcock and Fritted Disc
For flash chromatography
Supplier: Synthware™ C398030C
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
- With coarse porosity sealed in fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
| 73 mm | |
| 24/40 | |
| 305 mm |
| 1000 mL | |
| 4 mm bore |