Learn More
Synthware™ Chromatography Column with Standard Taper Joint, Reservoir, PTFE Stopcock and Fritted Disc
For flash chromatography
Supplier: Synthware™ C392640C
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
- With coarse porosity sealed in fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
| 20 mm | |
| 24/40 | |
| 457 mm |
| 250 mL | |
| 2 mm bore |