Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir and PTFE stopcock
For flash chromatography
Supplier:  Synthware™ C383225
 View more versions of this product
Catalog No. 31500946
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
  • Heavy-wall design increases safety
Specifications
Specifications
26 mm
35/20
254 mm
500 mL
2 mm bore
Product Suggestions
Product Suggestions