Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir and PTFE stopcock
For flash chromatography
Supplier: Synthware™ C381730
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
- Heavy-wall design increases safety
Specifications
| 13.4 mm | |
| 35/20 | |
| 305 mm |
| 100 mL | |
| 2 mm bore |