Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir, Fritted Disc and PTFE Stopcock
For flash chromatography
Supplier: Synthware™ C382640C
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
- With coarse porosity fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
| 20 mm | |
| 35/20 | |
| 457 mm |
| 500 mL | |
| 2 mm bore |