Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir, Fritted Disc and PTFE Stopcock
For flash chromatography
Supplier:  Synthware™ C382620C
 View more versions of this product
Catalog No. 31500954
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
  • With coarse porosity fritted disc to support column packing
  • Heavy-wall design increases safety
Specifications
Specifications
20 mm
35/20
203 mm
250 mL
2 mm bore
Product Suggestions
Product Suggestions