Learn More
Synthware™ Chromatography Column with Spherical Joint, PTFE stopcock and Fritted Disc
General purpose chromatography column for flash chromatography
Supplier:  Synthware™ C195463C
 View more versions of this product
Catalog No. 31500936
Only null left
Explore available promotions
Description
Description
  • Equipped with PTFE stopcock and top 35/20 socket joint
  • With coarse porosity fritted disc to support packing material
  • Synthware columns designed with heavy-walls for increased safety
Specifications
Specifications
40 mm
35/20
305 mm
Spherical joint
2 mm bore
Product Suggestions
Product Suggestions