Learn More
Synthware™ Chromatography Column with Spherical Joint, PTFE stopcock and Fritted Disc
General purpose chromatography column for flash chromatography
Supplier: Synthware™ C195173C
Description
- Equipped with PTFE stopcock and top 35/20 socket joint
- With coarse porosity fritted disc to support packing material
- Synthware columns designed with heavy-walls for increased safety
Specifications
| 13.4 mm | |
| 35/20 | |
| 305 mm |
| Spherical joint | |
| 2 mm bore |