Learn More
Synthware™ Filter Tubes with Fritted Disc
Used to filter particulates from substance
Supplier: Synthware™ F351430C
Description
- Standard taper inner joint
- Sealed in glass fritted disc of listed porosity
- Hose connector allows connection to a vacuum source
Specifications
| Two 2 mm glass stopcocks | |
| Coarse | |
| 14/20 | |
| ∼30 mm (Frit O.D.) |