Learn More
Synthware™ Addition Funnels with High Vacuum Valve
GREENER_CHOICE
Used for filling containers
$603.00 - $664.64
Specifications
Material Glass
Product Type Addition Funnel
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Joint Number Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Joint Number Price Quantity & Availability  
31501566
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F551060
100 mL 14/20
Each for $603.00
Only null left
31501567
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F554200
250 mL 24/40
Each for $664.64
Only null left
Description
Description
  • Upper outer and lower inner standard taper joints of same size
  • Supplied with two 0-4 mm high vacuum valves with PTFE O-ring at tip and PTFE wiper to protect Viton O-ring on shaft
Specifications
Specifications
Glass
Addition Funnel