missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Synthetic Blend Gray Knit Gloves

Constructed from a blend of medium weight cotton and synthetic fiber.

$8.56 - $8.56

Specifications

Construction String Knit
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Gray
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-120-757A
View Documents
Fisherbrand™
35C500S
Small
Dozen of 12 Pair for $8.56
Case of 25 Dozen for $199.25
Only null left
 
19-120-757B
View Documents
Fisherbrand™
35C500M
Medium
Dozen of 12 Pair for $8.56
Case of 25 Dozen for $199.25
Only null left
 
19-120-757C
View Documents
Fisherbrand™
35C500L
Large
Dozen of 12 Pair for $8.56
Case of 25 Dozen for $199.25
Only null left
 
19-120-757D
View Documents
Fisherbrand™
35C500XL
X-Large
Dozen of 12 Pair for $8.56
Case of 25 Dozen for $199.25
Only null left
 
Description

Description

  • Medium weight
  • Ideal for protecting hands from abrasion
  • Seamless style provides comfort and reversibility
  • Gray color hides dirt, allowing longer wear
  • Color-coded hems denote size
  • Full-fingered style
  • Washable
Specifications

Specifications

String Knit
Industrial
Gray
Reusable
Ambidextrous
Powder Free
Non-sterile
Continuous Knit
Yes
Uncoated
ANSI A1
No Texture
Cotton
String Knit Gloves
Full
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'