missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Synthegel™ 3D hiPSC Matrix Kit, 10 mL

Catalog No. 02707166
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02707166 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02707166 Supplier Corning™ Supplier No. 354787

Powerful tool for the physiological formation of 3D human induced pluripotent stem cell (hiPSC) spheroids in vitro. hiPSC spheroids maintain high viability, pluripotency and maintain differentiation potential based on multiple biomarkers.

  • Kit consists of a vial of Synthegel 3D hiPSC peptide nanofiber solution and a vial of Synthegel X-Link solution
  • Requires a vial of lyophilized Synthegel 3D hiPSC Grow Mix
  • Matrix rigidity is tunable by altering the peptide concentration
  • Purified synthetic peptide in neutral pH
  • Cells are encapsulated in physiological mimicking conditions
  • Fast hydrogel formation (5 to 30 minutes)
  • Sol-gel transfer reversible by shearing force
  • Works with standard cell culture methodologies, growth factors and media
  • Allows growth and passaging of hiPSCs in 3D format
  • The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
  • Allows 3D embedded hiPSC cultures

Specifications

Certifications/Compliance ISO
Color Red
Content And Storage Store at 2 to 8°C
Culture Environment Ambient Temperature
Description Synthegel™ 3D hiPSC Matrix Kit
Detection Method 3D Cell Culture
Form Liquid
Product Type Extracellular Matrix
Sterility Sterile-filtered
Format Kit
For Use With (Application) 3D embedded culture of hiPSC in a synthetic peptide hydrogel
pH Neutral
Product Line Hydrogel
Quantity 10 mL
Shipping Condition Ambient Temperature
Source Synthetic Peptide Matrix
Show More Show Less