missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Syntenin 1 Recombinant Rabbit Monoclonal Antibody (23GB2185)

Catalog No. MA553245
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553245 20 μL
MA553244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553245 Supplier Invitrogen™ Supplier No. MA553245
Only null left

Rabbit Recombinant Monoclonal Antibody

Syntenin 1 Recombinant Monoclonal Antibody for Flow, ICC/IF, IP, Western Blot

The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen Syntenin 1
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2185
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SDCBP
Gene Accession No. O00560
Gene Alias MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349)
Gene Symbols SDCBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human Syntenin (2-79AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6386
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less