Learn More
Syngene™ Gene Directory Track Database software
Syngene™ Gene Directory Track Database software with innovative design to give the user an array of configuration choices.
Supplier: Syngene™ GENEDIRECTORYDB
Description
- Used in conjunction with the GeneTools gel analysis package.
- Track classification with user defined fields.
- Genotyping capabilities.
- Powerful search facility for rapid identification of unknown tracks.
- Production of cluster analysis diagrams based on user defined parameters.
- VNTR table instantly calculated for any data set.
- Similarity matrix.
- Band matching to single or composite tracks.
- Dominance/Codominance marker identification with interface to JoinMap.
- GeneDirectory results create cluster analysis diagrams.
- Band match across multiple gels.
- Set up libraries with classification data.
- Make comparative quantifications.
- Share data between collaborating laboratories.
- Full report generation facility
Specifications
| Syngene Gbox | |
| Software Track Database |