missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader, IVD
GREENER_CHOICE

Catalog No. btn2abtsi
Change view
Click to view available options
Model:
N2MABT
NEO2
NEO2ALPHABT
NEO2ALPHAT
NEO2M
NEO2MALPHA
NEO2MALPHAB
NEO2MALPHAT
NEO2MB
NEO2MBT
NEO2MON
NEO2MT
NEO2S
NEO2SALPHA
NEO2SM
NEO2SMALPHA
NEO2SMALPHAB
NEO2SMB
NEO2T
Includes:
Monochromater: ABS
Monochromater: ABS, FI and LUM
Monochromater: ABS, FI and LUM; Alpha Laser
Monochromater: ABS, FI and LUM; TRF Laser
Monochromater: ABS; Alpha Laser
Monochromater: ABS; TRF Laser
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTN2ABTSI NEO2ALPHABT Monochromater: ABS
BTN2SI NEO2 Monochromater: ABS
BTN2SSI NEO2S Monochromater: ABS
BTN2TSI NEO2T Monochromater: ABS; TRF Laser
BTN2ATSI NEO2ALPHAT Monochromater: ABS
BTN2SASI NEO2SALPHA Monochromater: ABS; Alpha Laser
BTN2MSI NEO2M Monochromater: ABS, FI and LUM
BTN2MTSI NEO2MT Monochromater: ABS, FI and LUM; TRF Laser
BTN2MASI NEO2MALPHA Monochromater: ABS, FI and LUM; Alpha Laser
BTN2MABSI NEO2MALPHAB Monochromater: ABS, FI and LUM; Alpha Laser
BTN2MABTSI N2MABT Monochromater: ABS, FI and LUM; Alpha Laser
BTN2MATSI NEO2MALPHAT Monochromater: ABS, FI and LUM; Alpha Laser
BTN2MBSI NEO2MB Monochromater: ABS, FI and LUM
BTN2MBTSI NEO2MBT Monochromater: ABS, FI and LUM; TRF Laser
BTN2MONSI NEO2MON Monochromater: ABS, FI and LUM
BTN2SMSI NEO2SM Monochromater: ABS, FI and LUM
BTN2SMASI NEO2SMALPHA Monochromater: ABS, FI and LUM; Alpha Laser
BTN2SMABSI NEO2SMALPHAB Monochromater: ABS, FI and LUM; Alpha Laser
BTN2SMBSI NEO2SMB Monochromater: ABS, FI and LUM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. BTN2ABTSI Supplier Agilent BioTek Supplier No. N2ABTSI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For In Vitro Diagnostic Use.

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.

Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.

Specifications

Absorbance Range 230 to 999 nm, 1 nm increments
Accuracy Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
CO2 Concentration Range 0 to 20%
Depth (Metric) 52.5 cm
Depth (English) 20.7 in.
Incubator Temperature ±0.2°C at 37°C
Shaking Linear, Orbital, Double Orbital
Dynamic Range 0 to 4 OD
Dispense Volume 5 to 1000 μL, 1 μL increments
For Use With (Application) HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
Height (English) 16.1 in.
Height (Metric) 41 cm
Linearity ≤1% from 0 to 3 OD
Includes Monochromater: ABS
Model NEO2ALPHABT
Product Type Multi-Mode Microplate Reader
Power Consumption 250 W max.
Resolution 0.0001 OD
Weight (English) 78 lb.
Weight (Metric) 35 kg
Width (English) 15.4 in.
Width (Metric) 39 cm
Detector Type Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash
Show More Show Less