Learn More
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader, IVD

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek N2MBSI
Description
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.
Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.
Chemical Identifiers
| 230 to 999 nm, 1 nm increments | |
| CE and TUV marked. RoHS compliant. | |
| 52.5 cm | |
| ±0.2°C at 37°C | |
| 0 to 4 OD | |
| HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP | |
| 41 cm | |
| Multi-Mode Microplate Reader | |
| 0.0001 OD | |
| 35 kg | |
| 39 cm | |
| Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
| Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD | |
| 0 to 20% | |
| 20.7 in. | |
| Linear, Orbital, Double Orbital | |
| 5 to 1000 μL, 1 μL increments | |
| 16.1 in. | |
| ≤1% from 0 to 3 OD | |
| 250 W max. | |
| 78 lb. | |
| 15.4 in. | |
| Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option) | |
| Xenon Flash |
Specifications
| 230 to 999 nm, 1 nm increments | |
| CE and TUV marked. RoHS compliant. | |
| 52.5 cm | |
| ±0.2°C at 37°C | |
| 0 to 4 OD | |
| Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF | |
| 16.1 in. | |
| ≤1% from 0 to 3 OD | |
| NEO2MB | |
| 250 W max. | |
| 78 lb. | |
| 15.4 in. | |
| Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option) | |
| Xenon Flash |
| Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD | |
| 0 to 20% | |
| 20.7 in. | |
| Linear, Orbital, Double Orbital | |
| 5 to 1000 μL, 1 μL increments | |
| HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP | |
| 41 cm | |
| Monochromater: ABS, FI and LUM | |
| Multi-Mode Microplate Reader | |
| 0.0001 OD | |
| 35 kg | |
| 39 cm | |
| Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |