missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader
GREENER_CHOICE

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek N2MBTSN

 View more versions of this product

Catalog No. BTNEO2MBT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.

Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.

Specifications

Chemical Identifiers

230 to 999 nm, 1 nm increments
CE and TUV marked. RoHS compliant.
52.5 cm
±0.2°C at 37°C
0 to 4 OD
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
41 cm
250 W max.
78 lb.
15.4 in.
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Xenon Flash
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
0 to 20%
20.7 in.
Linear, Orbital, Double Orbital
5 to 1000 μL, 1 μL increments
16.1 in.
Multi-Mode Microplate Reader
0.0001 OD
35 kg
39 cm
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)

Specifications

230 to 999 nm, 1 nm increments
CE and TUV marked. RoHS compliant.
52.5 cm
±0.2°C at 37°C
0 to 4 OD
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
41 cm
NEO2MBT
250 W max.
78 lb.
15.4 in.
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Xenon Flash
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
0 to 20%
20.7 in.
Linear, Orbital, Double Orbital
5 to 1000 μL, 1 μL increments
16.1 in.
Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, TRF Laser
Multi-Mode Microplate Reader
0.0001 OD
35 kg
39 cm
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions