missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader, IVD
GREENER_CHOICE

Catalog No. BTSLXFASI
Change view
Click to view available options
Model:
SLXA
SLXATS
SLXF
SLXFA
SLXFATS
SLXFTS
Includes:
Absorbance
Absorbance, Fluorescence, Luminescence
Absorbance, Fluorescence, Luminescence, Touchscreen
Absorbance, Luminescence, Touchscreen
Fluorescence, Luminescence, Touchscreen
Fluorescence; Luminescence
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTSLXFASI SLXFA Absorbance, Fluorescence, Luminescence
BTSLXASI SLXA Absorbance
BTSLXATSS SLXATS Absorbance, Luminescence, Touchscreen
BTSLXFSI SLXF Fluorescence; Luminescence
BTSLXFATSSI SLXFATS Absorbance, Fluorescence, Luminescence, Touchscreen
BTSLXFTSSI SLXFTS Fluorescence, Luminescence, Touchscreen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTSLXFASI Supplier Agilent BioTek Supplier No. SLXFASI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek Synergy LX multi-mode reader automates many common microplate assays. For In Vitro Diagnostic Use.

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

Specifications

Absorbance Range 200 to 900 nm
Accuracy Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
Bandwidth ≤5 nm
Depth (Metric) 38.1 cm
Depth (English) 15 in.
Measurement Speed 24 sec. (96 wells), 76 sec. (384 wells)
Shaking Linear, Orbital, Double Orbital
Filters Monochromator Bandpass
For Use With (Application) Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
Height (English) 12 in.
Frequency 50/60 Hz
Height (Metric) 30.5 cm
Linearity <1% from 0 to 2.5 OD
Includes Absorbance, Fluorescence, Luminescence
Model SLXFA
Product Type Multi-Mode Microplate Reader
Power Consumption 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Resolution 0.001 OD
Sensitivity Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
USB Connections 2
Software Language Versions English
Warranty 1 year
Voltage 100/240 V
Weight (English) 27 lb.
Weight (Metric) 12.3 kg
Width (English) 15 in.
Width (Metric) 38.1 cm
Detector Type Photodiode, PMT
Software Type Full data analysis and reporting (under Gen5 control)
Light Source Xenon Flash
Reading Options End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Show More Show Less