Learn More
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader, IVD

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For In Vitro Diagnostic Use.
Specifications
| Absorbance Range | 200 to 999 nm, 1 nm increments |
|---|---|
| Accuracy | Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Depth (Metric) | 38 cm |
| Compatibility | BioStack and Third Party Automation Compatible |
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
BTS1ASI
![]() |
Agilent BioTek
S1ASI |
S1A | Absorbance, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTS1LSI
![]() |
Agilent BioTek
S1LSI |
S1L | Luminescence, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTS1LASI
![]() |
Agilent BioTek
S1LASI |
S1LA | Absorbance, Luminescence, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTS1LFASI
![]() |
Agilent BioTek
S1LFASI |
S1LFA | Absorbance, Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTS1LFSI
![]() |
Agilent BioTek
S1LFSI |
S1LF | Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTS1LFTASI
![]() |
Agilent BioTek
S1LFTASI |
S1LFTA | Absorbance, Luminescence, Fluorescence (Top/Bottom), Time-resolved Fluorescence (TRF - Secondary Mode), Alpha, Incubation, Shaking |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows
With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.
Specifications
| 200 to 999 nm, 1 nm increments | |
| CE and TUV marked. RoHS compliant. | |
| BioStack and Third Party Automation Compatible | |
| 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) | |
| Linear, Orbital | |
| 10 in. | |
| 25.4 cm | |
| Multi-Mode Microplate Reader | |
| 100/240 V | |
| 18 kg | |
| 40.6 cm | |
| Photodiode, PMT | |
| Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
| Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% | |
| 38 cm | |
| 15 in. | |
| 31 sec. (96 wells), 80 sec. (384 wells) | |
| Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades | |
| 50/60 Hz | |
| 2 Syringe Pumps | |
| 2 | |
| 40 lb. | |
| 16 in. | |
| 6- to 384-well Plates | |
| Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| End Point, Kinetic, Spectral Scanning, Well Area Scanning |