missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy HTX Multi-Mode Microplate Reader
GREENER_CHOICE

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek S1LSN

 View more versions of this product

Catalog No. BTS1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows

With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.

Order Info

Optional Accessories: Dual Reagent Dispenser Module, Gen5 Secure (for 21 CFR Part 11 Compliance), Fluorescence Test Plate, Absorbance Test Plate, Luminescence Test Plate, Patented BioCell 1 cm quartz vessel, Product Qualification Package, Take3/Take3 Trio.

Certifications

CE and TUV marked

Compliance

CE and TUV marked

Specifications

Chemical Identifiers

200 to 999 nm, 1 nm increments
CE and TUV marked. RoHS compliant.
BioStack and Third Party Automation Compatible
4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C)
Linear, Orbital
10 in.
25.4 cm
Multi-Mode Microplate Reader
100/240 V
18 kg
40.6 cm
Photodiode, PMT
Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity)
Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2%
38 cm
15 in.
31 sec. (96 wells), 80 sec. (384 wells)
Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades
50/60 Hz
2 Syringe Pumps
2
40 lb.
16 in.
6- to 384-well Plates
Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
End Point, Kinetic, Spectral Scanning, Well Area Scanning

Specifications

200 to 999 nm, 1 nm increments
CE and TUV marked. RoHS compliant.
BioStack and Third Party Automation Compatible
4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C)
Linear, Orbital
10 in.
25.4 cm
S1L
Multi-Mode Microplate Reader
100/240 V
18 kg
40.6 cm
Photodiode, PMT
Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity)
Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2%
38 cm
15 in.
31 sec. (96 wells), 80 sec. (384 wells)
Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades
50/60 Hz
Luminescence, Incubation, Shaking
2 Syringe Pumps
2
40 lb.
16 in.
6- to 384-well Plates
Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
End Point, Kinetic, Spectral Scanning, Well Area Scanning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions