Learn More
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For Research Use Only. Not for use in diagnostic procedures.
Supplier: Agilent BioTek S1ASN
Description
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows
With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.
Order Info
Optional Accessories: Dual Reagent Dispenser Module, Gen5 Secure (for 21 CFR Part 11 Compliance), Fluorescence Test Plate, Absorbance Test Plate, Luminescence Test Plate, Patented BioCell 1 cm quartz vessel, Product Qualification Package, Take3/Take3 Trio.
Certifications
CE and TUV marked
Compliance
CE and TUV marked
Chemical Identifiers
| 200 to 999 nm, 1 nm increments | |
| CE and TUV marked. RoHS compliant. | |
| BioStack and Third Party Automation Compatible | |
| 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) | |
| Linear, Orbital | |
| 10 in. | |
| 25.4 cm | |
| Multi-Mode Microplate Reader | |
| 100/240 V | |
| 18 kg | |
| 40.6 cm | |
| Photodiode, PMT | |
| Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
| Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% | |
| 38 cm | |
| 15 in. | |
| 31 sec. (96 wells), 80 sec. (384 wells) | |
| Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades | |
| 50/60 Hz | |
| 2 Syringe Pumps | |
| 2 | |
| 40 lb. | |
| 16 in. | |
| 6- to 384-well Plates | |
| Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| End Point, Kinetic, Spectral Scanning, Well Area Scanning |
Specifications
| 200 to 999 nm, 1 nm increments | |
| CE and TUV marked. RoHS compliant. | |
| BioStack and Third Party Automation Compatible | |
| 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) | |
| Linear, Orbital | |
| 10 in. | |
| 25.4 cm | |
| S1A | |
| Multi-Mode Microplate Reader | |
| 100/240 V | |
| 18 kg | |
| 40.6 cm | |
| Photodiode, PMT | |
| Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity) |
| Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% | |
| 38 cm | |
| 15 in. | |
| 31 sec. (96 wells), 80 sec. (384 wells) | |
| Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades | |
| 50/60 Hz | |
| Absorbance, Incubation, Shaking | |
| 2 Syringe Pumps | |
| 2 | |
| 40 lb. | |
| 16 in. | |
| 6- to 384-well Plates | |
| Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| End Point, Kinetic, Spectral Scanning, Well Area Scanning |