missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYNE3 Recombinant Rabbit Monoclonal Antibody (23GB4665)
GREENER_CHOICE

Catalog No. MA552726
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552726 100 μL
MA552727 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552726 Supplier Invitrogen™ Supplier No. MA552726
Only null left

Rabbit Recombinant Monoclonal Antibody

SYNE3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system. Plays a role in the regulation of aortic epithelial cell morphology, and is required for flow-induced centrosome polarization and directional migration in aortic endothelial cells. [UniProt].

Specifications

Antigen SYNE3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4665
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SYNE3
Gene Accession No. Q6ZMZ3
Gene Alias C14orf139; C14orf49; KASH domain-containing protein 3; KASH3; LINC00341; Nesp3; nesprin-3; NET53; Nuclear envelope spectrin repeat protein 3; RGD1304563; spectrin repeat containing nuclear envelope family member 3; spectrin repeat containing, nuclear envelope family member 3; SYNE3
Gene Symbols SYNE3
Host Species Rabbit
Immunogen A synthesized peptide derived from human Nesprin3 (750-935AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161176
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less