missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Synaptoporin Monoclonal Antibody (K49016_6D3), Invitrogen™

Catalog No. MA556024
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556024 200 μL
MA555644 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556024 Supplier Invitrogen™ Supplier No. MA556024
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: TGWHSSGQRY LSDPMEKHSS

Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).

Specifications

Antigen Synaptoporin
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K49016_6D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SYNPR
Gene Accession No. P22831, Q8TBG9
Gene Alias 1500003F20Rik; SPO; Synaptoporin; synaptorin; SYNPR
Gene Symbols SYNPR
Host Species Mouse
Immunogen Human SYNPR peptide (immunogen range is 201-220).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132204, 66030
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less