missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Synapsin II Recombinant Rabbit Monoclonal Antibody (23GB4960)

Catalog No. MA552745
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552745 20 μL
MA552744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552745 Supplier Invitrogen™ Supplier No. MA552745
Only null left

Rabbit Recombinant Monoclonal Antibody

Synapsin II Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.

Specifications

Antigen Synapsin II
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4960
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SYN2
Gene Accession No. Q63537, Q92777
Gene Alias 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb
Gene Symbols SYN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Synapsin II (210-270AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29179, 6854
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less