missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Synapsin 1 Recombinant Rabbit Monoclonal Antibody (3I9R10)
GREENER_CHOICE

Catalog No. MA558172
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA558172 100 μL
MA558171 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA558172 Supplier Invitrogen™ Supplier No. MA558172

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EDKQLIVELV VNKMAQALPR QRQRDASPGR GSHGQTPSPG ALPLGRQTSQ QPAGPPAQQR PPPQGGPPQP GPGPQRQGPP LQQRPPPQGQ QHLSGLGPPA.

SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen Synapsin 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 3I9R10
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SYN1
Gene Accession No. O88935, P09951
Gene Alias Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S
Gene Symbols SYN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 401-500 of human Synapsin-1 (NP_008881.2).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20964, 24949
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less