missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Swing Out Rotor
OHAUS™ Swing Out Rotors for use with the Frontier 5000 Series Multi Centrifuge
Supplier: OHAUS™ 30314824
Description
- Swing Out Rotors are available in a variety of capacities, and speeds
- For use with Frontier 5000 Series Multi Centrifuges
Chemical Identifiers
| Stainless Steel | |
| Frontier 5000 Series Multi Centrifuge | |
| Yes, when used with biocontainment lids |
| Swing-out Rotor | |
| 0 to 90° |
Specifications
| Rotor Swing out 2x3MTP w/ bucket ID | |
| Swing-out Rotor | |
| 2,716 x g | |
| 1.640 kg | |
| 0 to 90° |
| Stainless Steel | |
| 2 x 3 MTP | |
| 4,500 rpm | |
| Frontier 5000 Series Multi Centrifuge | |
| Yes, when used with biocontainment lids |