Learn More
3M™ Swab-Sampler, Sold by AquaPhoenix Scientific
Convenient and quick collection device for environmental sampling and testing over small surface areas
Supplier:  3M™ RS9604LET
 View more versions of this product
Catalog No. S26768
Only null left
Description
Description
  • Letheen broth vial ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Specifications
Specifications
Environmental sampling and testing over small surface areas
100/Cs.
4 mL letheen broth
Swab
Product Suggestions
Product Suggestions