Learn More
3M™ Swab-Sampler, Sold by AquaPhoenix Scientific
Convenient and quick collection device for environmental sampling and testing over small surface areas
$654.43 - $654.43
Specifications
| For Use With (Application) | Environmental sampling and testing over small surface areas |
|---|---|
| No. per Case | 100/Cs. |
| Product Type | Swab |
Description
- Letheen broth vial ready to use for pathogen testing of environmental samples
- Swab reaches into corners and hard-to-reach areas for increased accuracy
- Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
- Bottles have ample headspace for sample addition and vortexing
- Self-contained swab and vial for increased productivity and efficiency
Specifications
| Environmental sampling and testing over small surface areas | |
| 100/Cs. | |
| Swab |