missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SV2A Monoclonal Antibody (K70011_6B6), Invitrogen™

Catalog No. MA555664
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555664 50 μL
MA556044 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555664 Supplier Invitrogen™ Supplier No. MA555664
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: FIRGAKDIAK EVKKHAAKKV VKGLDRVQDE YSRRSYSRFE EEDDDDDFPA PSDGYYRGEG TQDEEEGGAS SDATEGHDED DEIYEGEYQG IPRAESGGKG ERMADGAPLA GVRGGLSDGE GPPGGRGEAQ RRKEREELAQ QYEAILRECG

This protein plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.It is identified as the brain binding-site for the antiepileptic drug levetiracetam/lev.

Specifications

Antigen SV2A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K70011_6B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SV2A
Gene Accession No. Q02563, Q7L0J3
Gene Alias AI746429; calcium regulator SV2A; Kiaa0736; mKIAA0736; PSEC0174; SV2; SV2A; synaptic vesicle glycoprotein 2 a; synaptic vesicle glycoprotein 2a; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA
Gene Symbols SV2A
Host Species Mouse
Immunogen Recombinant human SV2A (immunogen range is 11-160).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117559, 9900
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less