missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Electronic/Cleanroom Grade, 99.5%, Spectrum™ Chemical

Catalog No. 18611120
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611120 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18611120 Supplier Spectrum Chemical Mfg Cor Supplier No. I10571GL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Acidity 0.2 μeq/g
CAS Max % 1
Alkalinity 0.1 μeq/g
Quantity 1 gal.
Packaging Poly Bottle
Residue after Evaporation 5 ppm
Boiling Point 78.3°C
Percent Purity 99.5%
Grade Electronic/Cleanroom
Chemical Name or Material Isopropyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.