missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SureCast™ TEMED
Description
Experience superior performance, shelf life and convenience using Invitrogen™ SureCast™ polyacrylamide gel casting solutions.
SureCast TEMED (N,N,N',N'-tetramethylethylenediamine) is an accelerator for the polymerization of acrylamide and biacrylamide to prepare polyacrylamide gels for electrophoresis.
Recommended Storage
Room Temperature
Warranty
Up to 12 months
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Product Line | SureCast |
| Type | TEMED |
| Quantity | 30 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.