missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Glass Plates

Catalog No. HC1001
Encompass
Change view
Click to view available options
Quantity:
2 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC1001 2 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HC1001 Supplier Invitrogen™ Supplier No. HC1001
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.

SureCast Glass Plates offer superior quality, durability and damage-resistance and are used to cast 1.0mm polyacrylamide gels using the SureCast Handcast Station. SureCast gels are designed for use in the Mini Gel Tank. SureCast Gel Spacers not included and are sold separately.

Product Features
  • Premium, ultra-durable glass plates
  • Damage-resistant design
  • Cast up to 20 times more gels using a set of glass plates versus main competitor's glass plates
  • Compatible only with the SureCast Handcast System

Recommended Storage

Room Temperature

Warranty

Up to 12 months

Specifications

Content And Storage 2 front plates and 2 back plates
For Use With (Equipment) SureCast Handcast System
Gel Size Mini
Product Line SureCast
Type SureCast™ Glass Plates
Quantity 2 sets
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.