missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SureCast™ Gel Spacer
Description
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
The SureCast Gel Spacers are a replacement set of sealing gaskets for use with the SureCast Glass Plates to create a leak-free cassette used in the SureCast Handcast Station. Comes in a package of 10 gel spacers.
- Compatible only with the SureCast Handcast System
Recommended Storage
Room Temperature
Warranty
Up to 12 months
Specifications
Specifications
| For Use With (Equipment) | SureCast Glass Plate |
| Gel Size | Mini |
| Product Line | SureCast |
| Type | SureCast™ Gel Spacer |
| Quantity | 10 spacers |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.