missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Gel Handcast Station

Catalog No. HC1000
Encompass
Change view
Click to view available options
Quantity:
1 station
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC1000 1 station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HC1000 Supplier Invitrogen™ Supplier No. HC1000
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Leak-free, confident handcasting of your polyacrylamide gels with the SureCast Handcast System.

Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.

Pour your own polyacrylamide gels easily and confidently with the SureCast Handcast System and SureCast Handcast Reagents. The system features durable glass plates that are securely sealed using a single motion load-and-lock mechanism. The unique tilt feature of the SureCast Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.

Recommended Storage

Room Temperature

Warranty

Up to 12 months

Specifications

For Use With (Equipment) SureCast Handcast System
Gel Size Mini
Product Line SureCast
Type SureCast Gel Handcast Station
Quantity 1 station
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.